Universal Diamond ATR
Supplier: PerkinElmer
L1600129
470203-808EA 10746.27 USD
470203-808
Universal Diamond ATR
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Ensures Highly Reproducible Results
This universal ATR is an excellent general accessory for FT-IR analysis of solids and liquids using any spectrometer. Setup is simple with plug-and-play operation that requires no alignment and live force readout as samples are positioned. Produce high quality spectra through the use of a pressure arm allowing good contact of the sample with the diamond crystal. The pressure arm force indicator ensures first-class sample-to-sample and operator-to-operator reproducibility. A secure vapor-sealed lid ensures samples are protected from contamination, for accurate results with every use. Not for sale in Canada.