Quartz SUPRASIL Cuvettes
Supplier: PerkinElmer
B0631079
470203-800EA 813.79 USD
470203-800
Quartz SUPRASIL Cuvettes
Cuvettes Spectrophotometer Cuvettes
For Use with High Quality Spectrophotometer Units.
  • Macro Cuvette light path: 5 mm
  • Semi- and Ultra Micro Cuvette light path: 10 mm
These cells ensure top optical performance, with high cell transmission and accurate path length. The cell windows are fused to the bodies by a special process which avoids deformation of the windows. This assures that the full width of the cell is usable and the finished cell dimensions are maintained.
Ordering information: Not available in Canada.