Rolling Chemical Table
Supplier: Labconco
8020000
470203-268EA 1004.99 USD
470203-268
Rolling Chemical Table
Furniture Tables and Workbenches
For transporting laboratory supplies and equipment, table is made of 1" square epoxy-coated steel tubing and has two shelves of high pressure melamine laminate.
  • Rails around shelves
  • 4" hard rubber casters minimize chance of accidents
Shipping weight: 44 lbs.