LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Supplier: LaMotte
5860-01
470203-112EA 83.09 USD
470203-112
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range: 0 to 10 ppm
  • Materials for 50 tests
  • Aligned with GLOBE protocols
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.