LaMotte® Plankton Net
Supplier: LaMotte
1063
470203-064EA 142.47 USD
470203-064
LaMotte® Plankton Net
Specimens Live Specimens Aquaria - Living Material
Use this cone-shaped fine-mesh nylon net to let water pass while you filter out plankton.
  • 10 mesh, 153 micron nylon cloth
  • Clear, conical tube attached at end of the net allows plankon viewing
  • Net mouth braced by sturdy stainless steel ring and harness
A brass ring and rope harness keep the throat of the net open while the net is being towed. A conical, graduated tube (50 ml capacity) is secured to the end of the net for sample collection.
Delivery information: Includes spare graduated tube.