Fine-Tip Polyethylene Pipettes, Thermo Scientific
Supplier: Avantor
14670-325
470203-060CS 546.64 USD
470203-060
Fine-Tip Polyethylene Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Perfect for delivering small amounts of liquid.
  • Length 4"
  • Pipette draw 1.0 ml
  • Delivers 20 µl per drop