Visorgogs® Safety Goggles, Jones FR
Supplier: Jones & Co.
34862-RED
470202-894EA 19.4 USD
470202-894
Visorgogs® Safety Goggles, Jones FR
Goggles
These full-vision safety goggles protect eyes from flying particles or splashing liquids.
  • Indirect ventilation
  • Impact resistant
  • Opaque visor shades eyes and reduces eye strain
  • Full vision
  • Two visor colors available
The unique design incorporates comfortable, impact-resistant goggles with a vented, opaque visor to shade eyes and reduce eye strain and fatigue. The indirect ventilation screen keeps dirt and dust out and keeps lenses fog free. The visors are available in two colors.
The strutless lenses have a down-angle vision for clear visibility. Users can wear the goggles over prescriptions eyeglasses.
Certifications: Meets ANSI Z87+ standard.