Burner Wing Top
Supplier: Humboldt Manufacturing
H-6650 C-1505/1 H-6650
18026-108EA 18.88 USD
18026-108 470148-920 470202-860
Burner Wing Top
Heaters Gas Burners
It provides a broad, flat flame and fits mixing tubes with 1/2" outer diameter without stabilizer or 11/16" with stabilizer.