Triangular Support Stand
Supplier: Humboldt Manufacturing
H-21320
470202-852EA 44 USD
470202-852
Triangular Support Stand
Support Stands
Bases are cast iron with black enamel finish. Rods are zinc plated steel and threaded to fit into the base.
  • Base legs: 5"
  • Rod: 1/2" x 24"
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.