Aspirator Filter Pump
Supplier: Humboldt Manufacturing
H-12020
470202-842EA 30.45 USD
470202-842
Aspirator Filter Pump
Pumps Vacuum Aspiration Systems
Ideal for vacuum filtrations under both high and low water pressure.
  • A ball-type, positive action valve in the suction tube prevents backflow if pressure is reduced
  • Nickel-plated and designed for greater air entrainment capacity
  • Chapman, Swirling-type
  • 3/8" NPT tapered
  • Size: 5.375" L