Hemacytometer Set
Supplier: Hausser
3120
470202-800EA 493.2 USD
470202-800
Hemacytometer Set
Slides Counting Chambers
Obtain Accurate, Repeatable Blood Cell Counts
Features sharp contrast for lines and cells. Central square is ruled into 25 groups of sixteen squares, with each group separated by triple lines. The chamber is made of one-piece thermal and shock-resistant glass, with double Neubauer rulings in the two counting areas. The rulings patterns are 1/400 mm[2] and cover 9 mm[2]. Comes with counting chamber, two diluting pipets (RBC and WBC), two cover glasses, and two rubber tubing aspirators with mouthpieces. This set is supplied in a polyethylene case.