ExStik® pH100 Meter
Supplier: Extech
PH100 PH115
470202-636EA 246.15 USD
470202-636 470202-642
ExStik® pH100 Meter
Electrochemistry Meters pH and Multi-Parameter Meters
Flat electrode surface allows pH readings in liquid flms, semi-solids, and solids.
  • Aligned with GLOBE protocols
The meter itself has a smart chip that predicts electrode performance and has a RENEW indicator light when a replacement is needed. The CAL indicator will alert the user to recalibrate and users can perform one, two, or three-point calibrations. Other features include a 15 reading memory, automatic temperature compensation, data validation indicators to show when the reading has stabilized, and a digital display with bar graph andMore Product Information