3,000 - Microscope Slide Cabinet
Supplier: Eberbach
E2840.00
470202-564EA 4169.1 USD
470202-564
3,000 - Microscope Slide Cabinet
Slide Holders
Keep your slides protected and housed in this attractive cabinet.
  • Large capacity allows you to have space for your growing collection
  • Keep slides organized and contained
There’s no need to sacrifice attractive styling or proper storage methods when keeping large numbers of microscope slides. This classic-design hardwood case is finished in attractive light oak and opens to reveal storage trays for thousands of slides, plus a convenient pullout drawer to hold a card index. Sturdy, aluminum slide trays are compartmented for laying slides flat, the recommended storage method for keeping specimens intact. Doors are supported by full-length hinges for a close fit.
Note: Each compartment holds two slides, doubling the capacity of your cabinet to 6,000 as your collection expands.