Acrylic PMP/PE Stopcock, Geissler Burets
Supplier: Dynalon
564715-0050
470202-530EA 207.85 USD
470202-530
Acrylic PMP/PE Stopcock, Geissler Burets
Burets
Inexpensive, break-resistant burets ideal for school use.
Each clear acrylic buret is furnished with a polymethylpentene stopcock body, polyethylene stopcock key, nitrile rubber sealing rings, and a polypropylene tip. The burets are individually calibrated to conform to B.S. 846 Class B.
Designed for use with acids and bases; exposure to alcohol and organic solvents should be avoided.