Portable Pipet-Aid
Supplier: Drummond
4-000-100
470202-498EA 540 USD
470202-498
Portable Pipet-Aid
Pipettes Pipette Controllers
Capable of up to eight hours of continuous use, the Portable Pipet-Aid can dispense liquids over 2000 times without recharging.
  • Device accepts plastic or glass pipettes from 1 to 100 ml
  • Lightweight
  • Battery powered
  • Filtered nosepiece
  • Eight hours of continuous use
The filtered, failsafe nosepiece provides positive protection for the electronic components and prevents cross-contamination due to over pipetting. A tissue-culture nosepiece also provides quick disconnect and easy assembly.
The cordless device features power built into the handle, so it can be used while recharging. A 9 V rechargeable NiMH battery powers the device.
Certifications: UL listed. CSA approved.
Ordering information: This unit includes a holster, wall bracket, 110 V recharger, and four replacement filters included.