Disposable UV Cuvettes, Brandtech Scientific INC.
Supplier: Avantor
47744-640
470201-928PK 178.9 USD
470201-928
Disposable UV Cuvettes, Brandtech Scientific INC.
Cuvettes Electroporation Cuvettes
Economical option for UV cuvettes.
  • Package of 100
Eliminate the need for cleaning, and the associated risk of sample carry-over and cross-contamination. These disposable UV cuvettes replace expensive, fragile quartz cuvettes and are excellent for DNA, RNA, and protein analysis between 230–900nm. Made from polycyclical olefin, they can be used with most popular solvents, acids, and alkaline solutions.