Petri Dish Spreaders, Bellco Glass
Supplier: Bellco Glass
565400090
470201-816CS 345.15 USD
470201-816
Petri Dish Spreaders, Bellco Glass
Spreaders Cell Spreaders
Ideal for Spreading Inoculum over Agar face in Petri dish Cultures.
Manufactured from 4 mm O.D. borosilicate glass rod.