Polypropylene Micro Funnel, Bel-Art
Supplier: Bel-Art
F14685-0035_SE
470201-804CS 53.15 USD
470201-804
Polypropylene Micro Funnel, Bel-Art
Funnels Filtration Funnels Micro Filter Funnels
Ideal for filling burets and other narrow laboratory apparatus. The top diameter (ID) is 35 mm and stem diameter is 8 mm. Package of 12.