Desiccator Cabinet
Supplier: Bel-Art
F42046-0000_SE
470201-788EA 36.95 USD
470201-788
Desiccator Cabinet
Desiccators Desiccator Cabinets
Monitor Window Shows Blue When Recharged, Pink When Saturated.
  • Latches for secure closure
  • Single perforated shelf
Desiccant cartridges may be placed underneath shelf. Door is hinged on the left with the latch on the right. The unit may be used with a reusable cartridge, available separately, that effectively absorbs moisture through perforations in its aluminum case. Can be regenerated in a 150°C/ 300°F oven in about three hours.