Dialysis Bag Holder
Supplier: Bel-Art
F18237-0000_SE
470201-746CS 222.05 USD
470201-746
Dialysis Bag Holder
Clamps General Purpose Clamps
Easy to attach.
  • The autoclavable clip attaches to the side of a beaker
  • Size 2" L x 0.625" W
  • Package of six