Polypropylene Buret Funnels, Bel-Art
Supplier: Bel-Art
F14685-0035_SE
470201-738PK 53.15 USD
470201-738
Polypropylene Buret Funnels, Bel-Art
Funnels Filtration Funnels Filter Funnels
With 60° angle. For filling burets, manometers, and more.
Top diameter: 35 mm. Stem diameter: 8mm. Stem lenght: 20mm.
Package of 12.