Thermolyne® Hotplate
Supplier: Thermo Fisher Scientific
HPA1915BQ
470201-608EA 547.75 USD
470201-608
Thermolyne® Hotplate
Heaters Hot Plates
Perfect for Small Laboratories.
  • Temperature range: 100–700°F
  • Size: 6 1/2"L x 7 1/2"W x 4 1/2"H
Thermostatic temperature control, embedded heating elements, plus an aluminum top plate combine provide excellent surface temperature stability and uniformity over the entire area.