High-Capacity Deionizer Cartridge
Supplier: Thermo Fisher Scientific
D8901
470201-592EA 206 USD
470201-592
High-Capacity Deionizer Cartridge
Water Purification Systems Deionizers
Great for lab use.
  • Equipped with hose nipples on both ends for connection with ordinary laboratory tubing
  • When spent, the color layer changes from bluish purple to brown
  • Uses ⅜" tubing
  • Ion exchange capacity: 1725 grains (as NaCl)
Avoid elaborate and expensive demineralizer setups to produce water for general use with this miniature multi-bed system that requires no additional equipment.