Liquinox® Critical Cleaning Liquid Detergent
Supplier: Alconox
1232
470201-488CS 354.95 USD
470201-488
Liquinox® Critical Cleaning Liquid Detergent
Cleaning Agents Detergents and Surface Cleansers
Liquinox is a concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water.
  • Replaces corrosive acids and hazardous solvents
  • Phosphate free, biodegradable and readily disposable
  • Free rinsing to give you reliable results and no interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100
It can be used to clean Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors. Passes inhibitory residue test for water analysis.
And it can be used to remove soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemical and solvents.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration. Corrosion testing may be advisable.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace.
Certifications: USDA authorized. FDA certified.
Caution: Will foam - not for spray or machine use.