40-Place Test Tube Support
Supplier: Accurate Wirecraft
220400ZN
470201-452CS 561.2 USD
470201-452
40-Place Test Tube Support
Racks Tube Racks
Welded steel, zinc plated. Consists of two shelf levels for guides and a third elevated bottom shelf on which the tubes rest. 40 positions, in four rows of 10 holes each, accept tubes up to 22 mm diameter. Dimensions: 3-1/8" H x 10-5/8" W x 4-3/4" D.