Test Tube Basket with Lid
Supplier: Ward's Science
180020 180020
470149-014CS 142.4 USD
470149-014 470201-100
Test Tube Basket with Lid
Racks Tube Racks
This corrosion-proof polypropylene basket is ideal for rinsing glassware — simply slide the lid on, rinse, and invert for easy draining. The basket holds test tubes for storage, sterilization, or incubation. Autoclavable to 121°C. Size: 17 x 17 x 16cm.