Burner Tripod Stand with Concentric Rings
Supplier: United Scientific Supplies
TCR6X9
470201-092EA 32.85 USD
470201-092
Burner Tripod Stand with Concentric Rings
Tripods
Have Exactly the Right Size Tripod for Every Job
  • Two concentric rings, innermost removable
  • Legs removable for easy storage
Cast iron ring with black enamel finish. Detachable zinc-plated legs are interchangeable for use with various sizes of rings.