Alconox® Detergent Powder
Supplier: Alconox
1104
470200-838CS 507.6 USD
470200-838
Alconox® Detergent Powder
Cleaning Agents Detergents and Surface Cleansers
Scientifically Formulated for Manual Cleaning.
  • Mild (pH 9-9.5) anionic powder detergent
  • Biodegradable
Performs equally well in hard or soft, cold or hot water. It leaves no residue after rinsing. Everything cleaned becomes bright, shiny and sparkling.