Slotted Weights, Set of 8
Supplier: VWR International
PH0037F-SS
470200-834EA 147.8 USD
470200-834
Slotted Weights, Set of 8
Weights Calibration Weights
Stainless Steel Slotted Masses.
  • Accurate machined masses
  • Includes plastic and metal rack with handle.
  • 8 masses totaling 1100g
Set of 8: (1) x 10 g; (2) x 20 g; (1) x 50 g; (1) x 100 g; (2) x 200 g; and (1) x 500 g.