Hooked Masses, Set of 9
Supplier: Troemner
30390785
470200-626KT 472.25 USD
470200-626
Hooked Masses, Set of 9
Weights Calibration Weights
Hooked Mass Sets provide accurate results.
  • Stainless Steel Construction
  • Accurate Results
  • Lathe-Turned for Maximum Precision
  • Plastic Storage Block Included
These hook weights are made of stainless steel.
The weights are lathe turned for maximum precision.
Ordering information: This set includes a plastic storage block.
Packaging: One set includes nine weights.