High-Low Alcohol Burner Stand
Supplier: VWR International
CH10654
470200-546CS 1452 USD
470200-546
High-Low Alcohol Burner Stand
Tripods
Perfect for any style alcohol lamp, this stand can be used in two different height levels (9 or 13 cm) by simply removing the wire gauze top, turning the stand on its side, and putting the top back on. It is constructed of stainless steel rod with an 8 x 10 cm wire gauze surface. The edges of the wire gauze are turned under for safety.