Neutralization by the Drop Titration Method Kit
Supplier: Kemtec
4-114
470200-526KT 181.85 USD
470200-526
Neutralization by the Drop Titration Method Kit
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
The Neutralization by the Drop Titration Method Kit contains an easy and hands-on titration experiment.
  • Ideal for Grades 8-12
  • Easy-To-Use
  • Good Fit for Students with Limited Lab Background
  • No Preparation Required
  • Necessary Materials Included
It's a great way to teach the principles of acid and base titration to students with a limited laboratory background. It eliminates the need for expensive laboratory equipment for teaching titration and includes all necessary materials, alleviating extensive lab preparation.
The kit is ideal for leading discussions about titrations, acids, bases, pH, buffers, or environmental or physiological buffering systems in a range of science applications from biology to environmental sciences to physiology.
Ordering information: This kit includes a SDS universal indicator, sodium hydroxide, hydrochloric acid, phenolphthalein indicator, dropper bottles, toothpicks, a universal indicator chart, and depression plates.