DNA Staining Kit
Supplier: LAB-AIDS INC (KITS) SE
170
470200-500KT
199
USD
470200-500
DNA Staining Kit
Stains
Electrophoresis Stains
Using the Feulgen procedure, students macerate, stain and mount specially grown and selected root tips included in the kit. Then, using a microscope, they examine and draw what they observe of the various stages of cell division.
Includes: 1 vial specially selected root tips; 2 drop controlled btls. each 1M hydrochloric acid, leucofuchsin dye, and sodium bisulfite solution; 1 drop controlled btl. dilute acetic acid; 30 plastic treating dishes; 30 transfer brushes; 30 microscope slides; 30 cover slips; 30 students worksheets and guides; 1 instruction manual with MSDS.