PYREX® West Condenser
Supplier: Corning
2700-200
470200-312EA 179.45 USD
470200-312
PYREX® West Condenser
Condensers
This borosilicate condenser is designed with a heavy walled outer jacket to provide long life. The lighter walled inner tube provides efficient heat transfer. To reduce breakage, tabulations are located on the same side.