Hexagel™ Electrophoresis Chamber, Edvotek
Supplier: Edvotek
680-B 684-B 515
470205-942EA 47.25 USD
470205-942 470120-872 470120-876 470200-300
Hexagel™ Electrophoresis Chamber, Edvotek
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Run six gels simultaneously.
  • Lifetime warranty
  • Contoured lid for enhanced gel visualization
  • Large color coded push tabs for easy lid insertion and removal
  • Pour spout for buffer disposal
  • User replaceable electrodes
Apparatus supports up to six groups of students. Produces excellent results in 30 to 40 minutes.
Ordering information: Includes: (6) 7×7" Gel Trays; (6) 6-tooth combs; and (12) Rubber end dams.