Volta's Hail Storm
Supplier: Science First
N-132
470003-444EA 82 USD
470003-444 470200-214
Volta's Hail Storm
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A Perfect accessory for the Electrostatic Generator
  • Simple to use
  • Attaches to any Van De Graaff
Place pith balls or bits of aluminum foil in this cylinder to see how particles are attracted to the upper plate by induction, then repelled upon contact.