4 Decade Resistance Box
Supplier: Wards
PH1133A4
470200-200EA 99.7 USD
470200-200
4 Decade Resistance Box
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The 4 decade resistance box is a handy and convenient way to investigate resistance.
It provides four decade ranges from 0 to 11110W. Each decade is selected by a rotary switch.
All resistance values are ± 5% with a rating of 2W maximum per range. Two tapped points are provided for output.