Compact Sling Psychrometers
Supplier: Bel-Art Products
B61503-0000 B61503-0100
470200-002EA 94.55 USD
470200-002 470204-658
Compact Sling Psychrometers
Hygrometers Psychrometers
Measure relative humidity with weather bureau-type wet and dry bulb thermometers. Pocket-sized psychrometer is perfect for student use, as it presents no mercury hazards.
  • Two organic liquid filled thermometers with white back glass are housed in a durable case
  • Available in both Farenheit and Celsius
  • Replacement wicks available