Economy Alcohol Burner
Supplier: C & A Scientific
97-5313
470199-936CS 782 USD
470199-936
Economy Alcohol Burner
Heaters Spirit Lamps
Tip-resistant ethyl alcohol burner.
  • Nickel-plated snuffer cap extinguishes the flame and seals to prevent evaporation
  • Designed to burn 95% Denatured Ethyl Alcohol
Ordering information: Includes one wick. Alcohol is not included.