Ward's® Live Proteus vulgaris Culture - PATHOGEN
Supplier: Avantor
851172
470179-538KT 34.95 USD
470179-538 470198-316
Ward's® Live Proteus vulgaris Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Isolated from human urinary tract and wound infections, this organism is identifiable by its characteristic putrefactive odor. 
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism is pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.