Ward's® Live Drosophila melanogaster - Chromosome II Mutants
Supplier: Ward's Science
CALBP CALDUMP CALVEBRN 876610 CALBRN CALBLACK CALBLKVES 876572 876565
470176-738EA 10.95 USD
470176-738 470176-764 470176-768 470176-770 470176-780 470180-420 470180-422 470180-444 470180-448 470194-738 470194-744
Ward's® Live Drosophila melanogaster - Chromosome II Mutants
Specimens Live Specimens Animals - Living Material
Drosophila melanogaster, the common fruit fly, is widely used in genetic studies. They express a number of mutations such as eye, wing, and body mutations.
Students can study sex-linked, recessive, and dominant mutations. Drosophila also make an excellent live food for mantises, small reptiles and amphibians. Drosophila demonstrates complete metamorphosis with egg, larva, pupa, and adult stages in its life cycle.
The pupae will usually appear within ten days of the ship date.
Ordering information: Please specify a delivery date during checkout.
Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
During very cold weather, adult flies may die but eggs and larvae usually survive. Allow cultures to come to room temperature before checking under low magnification for eggs and live larvae.
Packaging: A large, vigorous culture is shipped in a shatterproof plastic vial.