Promega GoTaq® DNA Polymerase
Supplier: PROMEGA SUNNYVALE INC SE
M3001
470193-528EA 59 USD
470193-528
Promega GoTaq® DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
This Promega offers enhanced amplification over conventional Taq DNA polymerases. Supplied with 5X colorless reaction buffer and a 5X green buffer, which both allow for direct loading into agarose gels with no loading dye required. The included 5X green buffer allows you to visualize migration during electrophoresis. Each tube contains 100 U at 5 U/μl. Note: Requires storage at − 20°C.