Nutrient Agar
Supplier: IPM SCIENTIFIC, INC.
880004
470193-230EA 29.65 USD
470193-230
Nutrient Agar
Media for Microbiology Dehydrated Media for Microbiology
Multipurpose media. Supports the growth of a variety of microorganisms.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.