Trypic Soy Agar, Dehydrated
Supplier: IPM SCIENTIFIC, INC.
880002
470193-226EA 29.7 USD
470193-226
Trypic Soy Agar, Dehydrated
Media for Microbiology Dehydrated Media for Microbiology
Soybean-casein digest medium supports the growth of a variety of microorganisms. Premeasured packs save time in the lab. Simply add the contents of one dehydrated media pack to one liter of water, bring to a boil and sterilize to make enough media for 40 to 50 plates.