Super Sand
Supplier: SILVER CIRCLE PRODUCTS SE
SDRB-462
470193-074LB 12.85 USD
470193-074
Super Sand
Chemicals for Science Education
Super sand is hydrophobic sand that repels water. It never gets wet-put it in water and take it back out, and super sand stays completely dry.
  • Comes in a 1 pound bag
  • Use it to make underwater sand sculptures, clean up oil, improve plant growth, and much more
Caution: Do not handle with uncovered hands as the oil on our hands will reverse the reaction.