(S)-(-)-Nicotine solution, Reagent Grade

Supplier: ALDON CORP SE
Warning


Packaged under nitrogen.

NN0195-100
470191-572EA 21.55 USD
470191-572
(S)-(-)-Nicotine solution, Reagent Grade
(S)-(-)-Nicotine

Cigarette Solution / Cigarette Water Extract


Composition:
Water > 99.00%
Sodium benzoate 0.25%
Nicotine
Tar Not known

Formula: C₁₀H₁₄N₂
MDL Number: MFCD00006369
CAS Number: 54-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR