Polypropylene Powder Funnels
Supplier: VWR International
470191-320
470191-320CS 220.15 USD
470191-320
Polypropylene Powder Funnels
Funnels Powder Funnels
These unbreakable polypropylene powder funnels are ideal for student use. External ribs prevent air lock. Can be autoclaved. Stem length is 35 mm.