VWR® Wide-Mouth Flask
Supplier: VWR International
470191-168 470191-246 470191-250 470191-252
470191-168CS 376.75 USD
470191-168 470191-246 470191-250 470191-252
VWR® Wide-Mouth Flask
Flasks Erlenmeyer Flasks
High Quality, Great Value.
  • Borosilicate glass
  • Uniform wall thickness
  • White enamel metric scale
  • Marking spot
Each flask can be heated and used with acids, salt solutions, and organic solvents.