VWR® Cylinders with Plastic Base
Supplier: VWR International
470191-190 470191-192 470191-194 470191-206
470191-190CS 97.1 USD
470191-190 470191-192 470191-194 470191-206
VWR® Cylinders with Plastic Base
Cylinders Measuring Cylinders
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
  • Double graduated metric scale
  • Pour-out
  • Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.