Vernier® Chemical Polarimeter
Supplier: Vernier Software & Technology
CELLS-POL
470134-260EA 578.99 USD
470134-260 470189-708
Vernier® Chemical Polarimeter
Polarimeters
Measure chiral properties without destruction.
  • Students no longer have to determine the optical maximum by eye
The Polarimeter can be used to measure chiral properties of optically active samples without chemically modifying or destroying the sample. This is a vertical polarimeter that uses a 589 nm LED, a fixed polarizer, and a manually rotated polarizer to detect changes in rotation of plane-polarized light in the presence of an optically active compound. Used with Vernier technology, students no longer have to determine the optical maximum with their eye but have a graph that shows a clear change in the light’s polarization.
Sensitivity: typically up to OD 2
Optical Encoder: Bidirectional, quadrature encoder, 360 cycle per revolution
Light Source: LED
Wavelength (nominal): 589 nm
For educational use only.